SP Bel-Art Policeman Stirrer, Polypropylene, Bel-Art Products, a part of SP
# de catalogue 53800-005
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- Length:20.3 cm (8")
- Cat. no.:53800-005
- Blade Width:4.8 mm (³/₁₆")
- Supplier No.:377390001
Spécifications
A propos de cet article
This policeman stirrer is made of a 4.7 mm (³/₁₆") Ø polypropylene rod, with one end flattened and cut diagonally.
- Ergonomic design
- Polypropylene rod construction
- Suitable for scraping, transferring materials, and stirring
- Autoclavable
Useful for scraping and transferring materials, it is steam autoclavable at 121 °C (250 °F).
Customers Who Bought This Also Bought