LabFit™ Base Cabinet Drawers
Fournisseur: TrippNT
New Product
Certificats
A propos de cet article
TrippNT® LabFit™ casework is constructed of half-inch polypropylene which is ideal for highly corrosive enviroments.
- Non-metal construction
- Resistant to chemicals and corrosion
- Easy to clean and disinfect
- Large removeable rear access panel
- Industry leading two year full replacement warranty
- Made in USA
Polypropylene is a versatile plastic material that offers numberous beneftis for controlled environments. Its durability and resistance to corrosion make it a practical choice. It can endure high temperatures and exposure to various chemicals, making it an ideal option for a range of laboratory and medical applications. Joints are welded insuring a leak-tight, flush union. All hinges, handles, drawer slides, and stops are constructed of polypropylene.
Delivery: TrippNT® LabFit laboratory casework has all the components necessary for a complete laboratory set-up. For further information, contact your Avantor furniture specialist.
Customers Who Bought This Also Bought