Magnetic Probe Holder for Positioning the PT100 Probes Easily in Refrigerators, Freezers, and Other Assets
# de catalogue 77394-758
Fournisseur: SENSEANYWHERE BV
New Product
Spécifications
- Description:The magnetic probe holder is perfect for positioning the PT100 probes easily in refrigerators, freezers, and other assets, boasting user-friendly installation
Spécifications
A propos de cet article
The magnetic probe holder is perfect for positioning the Pt100 probes easily in refrigerators, freezers, and other assets, boasting user-friendly installation.
The hole diameter is precisely engineered between 3.1 to 3.2 mm to ensure a universal fit for both stainless steel and hermetically sealed probes, guaranteeing compatibility across a range of models.
Customers Who Bought This Also Bought