A propos de cet article
Use credits to easily register your devices, receive SMS alerts, receive voice alerts and obtain calibration certificates.
SenseAnywhere credits card: Prepaid subscription card with activation code for data loggers.
Top-half prepaid subscription card - this card represents two to 50 credits only valid for the SenseAnywhere portal. Use credits to easily register your devices, receive SMS alerts, receive voice alerts and obtain calibration certificates. For more details see help >credits once you have logged-in.
Bottom-half prepaid subscription card
1. Remove scratch-off paint from area above.
2. Go to the website listed.
3. Log-in or create a new account.
4. Follow instructions on the screen.
Credits are required to pay for usage of the SenseAnywhere portal, the storage of the data of your data loggers, sending SMS alerts and receiving voice alerts. If you want to send out alerts using text messages (SMS) take into account that 1/40 credits (0.025 credits) will be deducted from your credits balance per message. For telephoned Voice alerts 1/40 credits (0.025 credits) will be deducted per successful call attempt.
Credits will be deducted when:
A data logger is registered or its license is automatically renewed at the end of each subscription year.
For a voice notification, a successful connection is made in a calling list. This can be an accepted or rejected alert, or a connection to voicemail.
An SMS alert is sent: 1/40 credit (0.025 credit) per sent message.
A mobile access point with a SenseAnywhere SIM card is being registered or its mobile subscription is being renewed at the end of each subscription year.