ContactUsLinkComponent
Stilla® Sapphire Chip
Stilla® Sapphire Chip
# de catalogue 77392-264
New Product
Numéro CAS:  
SAPPHIRE CHIPS PK12
Stilla® Sapphire Chip
# de catalogue 77392-264
New Product
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Sapphire chip
  • Units:
    Pack of 12 chips
Spécifications
A propos de cet article
The Sapphire Chip features cutting-edge, patented microfluidic technology that integrates the entire digital PCR workflow into a single consumable.
  • Capacity: Up to four samples/chip
  • Input volume: 25 µl/chamber
  • Droplets per sample: Up to 30000
  • Dynamic range: ∼5 log
For maximum template input volume, the Sapphire Chip can be loaded with up to 4 samples per chip, and generate up to 30,000 droplets per sample for your digital PCR workflows. After sample preparation, just a single pipetting step is required before the chip is loaded onto either the 3-color naica® system or the 6-color naica® system for partitioning, PCR amplification, and sample read-out. The Sapphire Chip supports applications such as rare mutation detection, liquid biopsy analysis, gene expression analysis, and pathogen detection.
Customers Who Bought This Also Bought