About this item
The naica® mutliplex PCR MIX is a ready-to-use two-component solution specifically formulated for optimal TaqMan® fluorescent probe-based Crystal Digital PCR®.
- naica® mutliplex PCR MIX 5X - Available in 0.750 and 1.50 ml volume sizes
- naica® mutliplex PCR MIX 10X - Available in 0.375 and 0.750 ml volume sizes
The naica® multiplex PCR MIX was specially-formulated to ensure optimal performance for Taqman® fluorescent probe-based Crystal Digital PCR® on the Nio™+ and naica® systems. The stably incorporated internal blue channel reference dye facilitates reaction assembly while ensuring reliable partition detection.
The unique 10X concentration liberates a rection volume that can instead be utilized by additional sample input, probes, or primers, which increases the multiplexing capacity and provides superior detection sensitivity for low concentration samples and rare targets.