AdipoRed® Assay Reagent
# de catalogue CA10064-396
Fournisseur: LONZA PHARMA - BIOSCIENCE CA
Numéro CAS:
Spécifications
- Description:AdipoRed® Assay reagent
- Cat. no.:CA10064-396
- Supplier No.:PT-7009
Spécifications
A propos de cet article
The AdipoRed® Assay Reagent is designed for assessing the effect of compounds on the differentiation of preadipocytes or on lipid utilization in mature adipocytes. The lipophilic AdipoRed® Assay Reagent specifically partitions into the fat droplets of differentiated adipocytes and fluoresces at 572 nm.
- Convenient - Simply remove cell culture medium, add premixed reagent and read in a standard fluorometer
- Fast- Process an entire 96-well or 384-well plate in as little as 20 minutes
- Effective - Provides objective, high throughput measurement of the accumulation of intracellular triglycerides, with high signal to noise ratios
This objective, high-throughput, homogeneous fluorescence-based assay quantifies the accumulation of intracellular triglycerides and provides significant advantages to drug discovery efforts in the field of obesity and diabetes. It is more sensitive than other methods such as the Oil Red O assay, and is much faster and easier than Northern and Western blots.
Delivery: Content: 5 × AdipoRed® Assay Reagent, 4 ml.
Customers Who Bought This Also Bought