ContactUsLinkComponent
Bovine Brain Extract (BBE)
Bovine Brain Extract (BBE)
# de catalogue CA10063-928
Numéro CAS:  
Bovine Brain Extract, 9 mg/ml, 5 ml
Bovine Brain Extract (BBE)
# de catalogue CA10063-928
Numéro CAS:  
Spécifications
  • Cat. no.:
    CA10063-928
  • Supplier No.:
    CC-4098
Spécifications
A propos de cet article
Bovine Brain Extract is useful in the culture of various endothelial cell types. The product is offered at a concentration of 9 mg/ml.
Delivery: Content: 1 × Bottle containing 9 mg/ml Bovine Brain Extract, 5 ml.
Customers Who Bought This Also Bought