Bovine Brain Extract (BBE)
# de catalogue CA10063-928
Fournisseur: LONZA PHARMA - BIOSCIENCE CA
Numéro CAS:
Spécifications
- Cat. no.:CA10063-928
- Supplier No.:CC-4098
Spécifications
A propos de cet article
Bovine Brain Extract is useful in the culture of various endothelial cell types. The product is offered at a concentration of 9 mg/ml.
Delivery: Content: 1 × Bottle containing 9 mg/ml Bovine Brain Extract, 5 ml.
Customers Who Bought This Also Bought