Order Entry
ContactUsLinkComponent
Rat Brain Cortex Neurons
Rat Brain Cortex Neurons
# de catalogue CA10064-116
Numéro CAS:  
R-Cx - Rat Brain Cortex Neurons, Cryopreserved, in PNGM* BulletKit, =4 million cells/vial
Rat Brain Cortex Neurons
# de catalogue CA10064-116
Numéro CAS:  

Spécifications

  • Description:
    Rat brain cortex neurons
  • Size:
    ≥4 million cells
  • Cat. no.:
    CA10064-116

Spécifications

A propos de cet article

Ready-to-use Clonetics® Rat Brain Cortex Neurons from rat (E18, 19) brain are cell suspensions of high quality primary embryonic brain neuronal cells (including glia) prepared by standardized methods, and are ready for immediate culture.

  • Ready-To-Use
  • No tissue dissection or animal handling required
  • Increased data reproducibility
  • Characteristic morphology and staining tests confirm tissue origin and cell type
  • Negative for mycoplasma, bacteria, yeast and fungi

Each vial of cortical cells contains approximately 4 million viable cells. The cells will seed into 10 or more wells of a 24-well plate and 50 or more wells of a 96-well plate using the recommended plating densities and medium.Cell death will occur during the first few days after plating and debris will be observed. This is normal. After approximately 4 days in culture, the cells will form a neurite network and by the 7th day, debris will be minimal.

Recommended Medium: PNGM® BulletKit® (CC-4461)

Applications: Neurogenesis, Neurotransmitter function, Gene expression, Signaling pathways, Electrophysiology, Neurotoxicity, Drug/Compound screening, Advanced cell culture models.

Delivery: Content: 1 × Cryopreserved ampule of Rat Cortex Neurons containing ≥4000000 cells. Certificate of Analysis (CoA) is available for every batch.
Customers Who Bought This Also Bought