ContactUsLinkComponent
D-SAEC-As – Diseased Small Airway Epithelial Cells – Asthma
D-SAEC-As – Diseased Small Airway Epithelial Cells – Asthma
# de catalogue CA10064-336
Numéro CAS:  
Clonetics* D-SAEC-Asthma small airway epithelial cells cryoamp
D-SAEC-As – Diseased Small Airway Epithelial Cells – Asthma
# de catalogue CA10064-336
Numéro CAS:  
Spécifications
  • Description:
    D-SAEC-As – Diseased Small Airway Epithelial Cells – asthma
  • Size:
    ≥500000 cells
  • Cat. no.:
    CA10064-336
Spécifications
A propos de cet article
Asthma is an inflammatory disorder of the airways, which causes attacks of wheezing, shortness of breath, chest tightness, and coughing in patients. Degrees of severity are unique to individual patients as are responses to drug treatments.
Adhesion of eosinophils to bronchial epithelial cells, and the interaction of airway smooth muscle and sub-epithelial fibroblasts play a critical role in asthma pathology. During an asthma attack bronchiole smooth muscle tissue can constrict and decrease the flow of air in the airways. The amount of air flow can further be decreased by inflammation or excess mucus secretion.
Small airway epithelial cells stained for Cytokeratin 19. Human Small Airway Epithelial Cells (SAEC) are isolated from asthmatic human lung tissue the distal portion of the lung in the 1mm bronchiole area.
Applications:
Gene expression pathways.
Phenotypic changes to compounds.
Transfect diseased cells.
Co-culture with immune cells.
Airway remodeling studies.
Cellular interactions.
Cryopreserved SAECs from Asthma donors are guaranteed through 15 population doublings and stain positive for cytokeratin 19.All cells testnegative for mycoplasma, bacteria, yeast, and fungi. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots.
Delivery: Content: 1×cryopreserved Ampule with ≥500000 cells/vial and Certificate ofAnalysis is provided for each cell lot purchased.
Customers Who Bought This Also Bought