ContactUsLinkComponent
D-HMVEC-C – Diseased Human Cardiac Microvascular Endothelial Cells – Diabetes Type I
D-HMVEC-C – Diseased Human Cardiac Microvascular Endothelial Cells – Diabetes Type I
# de catalogue CA10064-326
Numéro CAS:  
Diseased Cardiac Microvascular Endothelial Cells Kit
D-HMVEC-C – Diseased Human Cardiac Microvascular Endothelial Cells – Diabetes Type I
# de catalogue CA10064-326
Numéro CAS:  
Spécifications
  • Description:
    D-HMVEC-C – Diseased Human Cardiac Microvascular Endothelial Cells – diabetes type I
  • Size:
    ≥500000 cells
  • Cat. no.:
    CA10064-326
  • Supplier No.:
    CC-2927
Spécifications
A propos de cet article
Diseased cardiac microvascular endothelial cells are isolated from human donors that have been diagnosed with diabetes type I or type II disease. Diseased cells are grown in the same optimized media system as normal human endothelial cells.
Diabetic cultures can be used for experimental applications in diabetic-related cardiovascular pharmaceutical development, vascular pathology, including atherosclerosis.
Cryopreserved Diabetic Cardiac Microvascular Endothelial Cells are shipped in third passage. Proliferating Clonetics® cells are shipped in fourth passage. All cells test negative for mycoplasma, bacteria, yeast, and fungi. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots.
Delivery: Content: 1×cryopreserved Ampule with ≥500000 cells/vial and Certificate of Analysis is provided for each cell lot purchased.
Customers Who Bought This Also Bought