OsteoImage® Mineralization Assay, 500 Test
# de catalogue CA10064-078
Fournisseur: LONZA PHARMA - BIOSCIENCE CA
Numéro CAS:
Spécifications
- Description:OsteoImage® Mineralization assay, 500 test
- Cat. no.:CA10064-078
- Supplier No.:PA-1503
Spécifications
A propos de cet article
The OsteoImage® mineralization assay is a rapid, fluorescent in vitro assay for assessing bone cell mineralization. The OsteoImage® assay can quantitate in vitro mineralization by osteogenic stem cells, primary osteoblasts, and osteoblast-like cell lines.
- Delivers qualitative visual fluorescent microscopy or quantitative fluorescent plate reader results
- Can be used with primary osteoblasts, osteoblast stem cells, and osteoblast cell lines
- Measures hydroxyapatite, similar to real bone
- Completed in less than 90 minutes, without tedious extractions
- Sensitive enough to detect time dependent increases in mineralization in differentiating cells
- Scalable for use in 6-well to 96-well plates
It is based on specific binding of the fluorescent OsteoImage® Staining Reagent to the hydroxyapatite portion of bone-like nodules deposited by cells.
Unlike typical histochemical methods such as von Kossa and Alizarin red, neither of which is hydroxyapatite specific, the OsteoImage® assay eliminates multiple steps or tedious extraction steps.
Increase the speed, sensitivity, and ease of measuring mineralization in your cell cultures with the OsteoImage® mineralization assay.
Delivery: Content:
5×96 well plates
1×500 μl OsteoImage® staining reagent
1×50 ml staining reagent dilution buffer
1×50 ml OsteoImage® wash buffer (10×)
5×96 well plates
1×500 μl OsteoImage® staining reagent
1×50 ml staining reagent dilution buffer
1×50 ml OsteoImage® wash buffer (10×)
Customers Who Bought This Also Bought