HPAEC – Human Pulmonary Artery Endothelial Cells
# de catalogue CA10064-210
Fournisseur: LONZA PHARMA - BIOSCIENCE CA
Numéro CAS:
Spécifications
- Description:HPAEC – Human Pulmonary Artery Endothelial cells
- Size:≥500000 cells
- Cat. no.:CA10064-210
- Supplier No.:CC-2530
Spécifications
A propos de cet article
Lonza's Human Pulmonary Artery Endothelial Cells (HPAEC) and media are convenient and easy to use, allowing the researcher to focus on results.
- Assured for experimental use up to 15 population doublings
- All cells test negative for mycoplasma, bacteria, yeast, and fungi
- HIV-1, Hepatitis B and Hepatitis C are not detected for all donors and/or cell lots
- Characterized by morphological observation throughout serial passage
Cryopreserved HPAEC cells are shipped in third passage. Proliferating cells are shipped in fourth passage. Cells isolated from Diabetes Type 1 and 2 are also available.
Clonetics® cells, medium and reagents are quality tested together and guaranteed to give optimum performance as a complete cell system.
Applications: Hypertension, arteriosclerosis, inflammation, oncology, drug screening.
Delivery: Content: 1×cryopreserved ampule of HPAEC containing ≥500000 cells.
Customers Who Bought This Also Bought