ContactUsLinkComponent
HMVEC-dNeo – Human Dermal Microvascular Endothelial Cells, Neonatal, Pooled
HMVEC-dNeo – Human Dermal Microvascular Endothelial Cells, Neonatal, Pooled
# de catalogue CA10064-204
Numéro CAS:  
EGM*-2MV BulletKit* HMVEC-dNeo - Human Dermal Microvascular Endothelial Cells, Neonatal, Pooled
HMVEC-dNeo – Human Dermal Microvascular Endothelial Cells, Neonatal, Pooled
# de catalogue CA10064-204
Numéro CAS:  
Spécifications
  • Description:
    HMVEC-dNeo – Human Dermal Microvascular Endothelial cells, neonatal, pooled
  • Size:
    ≥500000 cells
  • Cat. no.:
    CA10064-204
  • Supplier No.:
    CC-2516
Spécifications
A propos de cet article
Dermal microvascular endothelial cells include cells isolated from small vessels within skin tissue from either adults or neonatal foreskins.
Cells are cultured to be ≥90% pure and specifically enriched for dermal LECs and BECs. Dermal microvascular cells, from either neonates or adults (HMVEC-D) are a mixed population of both blood (HMVEC-DBl) and lymphatic (HMVEC-DLy) endothelial cells.
Clonetics® Cells, Medium and Reagents are quality tested together and guaranteed to give optimum performance as a complete cell system.
Cryopreserved cells are shipped in third passage. Cryopreserved Dermal Microvascular Endothelial Cells are guaranteed through 15 population doublings (CC-2543, CC-2505 & CC-2516) and 12 population doublings (CC-2810, CC-2811, CC-2812 & CC-2813) , express CD31/105, von Williebrand Factor VIII, and are positive for acetyated low density lipoprotein uptake. All cells test negative for mycoplasma, bacteria, yeast, and fungi. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots.
Delivery: Content: 1×cryopreserved ampule with ≥500000 cells/vial and Certificate of Analysis is provided for each cell lot purchased.
Customers Who Bought This Also Bought