Software for Lucetta® 2 Luminometer
# de catalogue 76321-330
Fournisseur: Lonza
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Description:Lucetta® connect software
- Cat. no.:76321-330
Spécifications
A propos de cet article
The Lucetta® connect software offers data transfer from the Lucetta® 2 luminometer to a PC for long-term storage. Measurement results can be viewed and analyzed in Microsoft® Excel®.
- Easy and intuitive handling
- Longterm storage and filing of assay results
- Creation of one Excel® File per assay
Delivery: Kit includes 1×USB stick with Lucetta® connect software.
Customers Who Bought This Also Bought