Eisco Heavy Wall Test Tubes
Fournisseur: Wards
New Product
Certificats
A propos de cet article
Durable heavy wall test tubes.
- Borosilicate glass 3.3, heavy wall, with rim
- Strong resistance to heat and chemicals
- 1.5 mm wall thickness
These products have a heavy wall with a rim. Has strong resistance to heat and chemicals. Comes in bulk packs. Comes in multiple different capacities.
Customers Who Bought This Also Bought