KIMBLE® Glass Wash Bottle, 1000 ml, DWK Life Sciences
# de catalogue KT1011
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Capacity:1000 ml
- Cat. no.:KT1011
- Supplier No.:1011
Spécifications
A propos de cet article
Unitized Wash bottles allow you to automatically pump a continuous, forceful jet of washing or rinsing liquid that may be instantly stopped as needed.
- Easy one-handed operation (either hand), leaving the other hand free for other needs
- Crystal clear glass flask permits quick inspection of the contents
- Unitized Wash bottles are recommended for use at ambient temperatures
- Features borosilicate glass flask and neoprene parts for improved solvent resistance
Customers Who Bought This Also Bought