PINK UNDERGUARD* Nitrile Examination Gloves, Powder-free, Latex-free
Fournisseur: O&M HALYARD, INC
New Product
Certificats
A propos de cet article
The first medical examination glove specifically designed to help healthcare professionals comply with recommended double gloving protocols when handling chemotherapy drugs.
- Coated exterior of the glove reduces friction
- Coating makes donning a second pair of gloves easy and alleviates the typical frustrations associated with double glovings
- Tested with 52 chemotherapy drugs in accordance with ASTM D6978
- Extended 12" cuff, providing additional coverage under the cuff of the chemo gown
- Pink color provides visible breach detection when wearing contrasting outer gloves
As new PPE protocols for handling chemotherapy agents are implemented, it is imperative to have a chemotherapy glove that appropriately protects healthcare professionals and makes compliance to double gloving easy. Powder-free PINK UNDERGUARD* nitrile examination gloves do both.
Spécifications
- Catalog No:
- 77322-592
- 77322-590
- 77322-596
- 77322-594
- 77322-598
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Material:
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Finish:
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile