Synthware High Vacuum Valve, Teflon® Protected, 3-Way Metering
# de catalogue 60004-040
Fournisseur: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- Description:High vacuum valve, Teflon® protected, 3-way metering
- Valve Size:0 - 8 mm
- Supplier No.:S640008
Spécifications
A propos de cet article
Viton® O-rings on PTFE shaft are protected with precision Teflon® wipers. The plug has a machined-in integral PTFE tip O-ring to make the primary seal.
- Teflon® shaft
- PTFE covered Viton® O-rings
Valves are designed to provide an excellent vacuum tight seal without exposing any material other than Teflon® to chemicals.
Customers Who Bought This Also Bought