Spécifications
- Description:StarterKit for open door monitoring in refrigerators and freezers, with AccessPoint, AssetTag and 5 credits
Spécifications
A propos de cet article
Discover the SenseAnywhere StarterKit with the AiroSensor 20-20-41 and Magnetic Door Contact Set for efficient open door monitoring in refrigerators and freezers. This kit includes an Indoor AccessPoint for secure data transfer, AssetTagging for easy sensor management, and five credits for optimal system functionality.
- The ultimate solution to your refrigeration and freezer challenges
- Program alarms for open or improperly closed doors
- Monitor door frequency and duration on refrigerators and freezers
- Easy mounting
The SenseAnywhere AiroSensor 20-20-41 with Magnetic Door Contact Set is the ultimate solution to your refrigeration and freezer challenges.
We all know the frustration of discovering that your fridge or freezer has failed due to an open door. With the AiroSensor 20-20-41 combined with the Magnetic Door Contact Set, you can say goodbye to this common problem.
With these two products, you can program alarms for open or improperly closed doors. This innovative solution monitors door frequency and duration on refrigerators and freezers to prevent potential disasters caused by open doors.
The connecter with cable can be easily shortened and the magnetic switch set with terminal screws can easily mount the cable, and they are provided with two double-sided nano-tape pads for easy mounting even in sub-zero temperatures.
Experience has shown that most alarms and excursions occur due to improperly closed doors. But with the AiroSensor 20-20-41 and Magnetic Door Contact Set, you can rest easy knowing that you are protected against these potential disasters.
Enjoy the peace of mind that comes with knowing your refrigeration and freezer systems are operating at peak performance. Don’t wait any longer - upgrade your systems today and join the thousands of satisfied customers who have already experienced the benefits of SenseAnywhere technology.
Indoor AccessPoint - The SenseAnywhere Indoor AccessPoint is the cornerstone of reliable and secure data transmission. It connects our Sensors to the SenseAnywhere Cloud Service, ensuring robust, two-way, fully encrypted communication. Sensors automatically detect the AccessPoint, seamlessly transmitting data to our central database. With advanced protocols for re-transmission and duplicate packet filtering, we guarantee that no event is ever missed. Designed specifically for indoor environments, this AccessPoint is an integral part of our robust surveillance solution.
AssetTagging - AssetTagging by SenseAnywhere revolutionizes sensor calibration and monitoring continuity. This system allows for uninterrupted data logging with interchangeable loggers, eliminating gaps during sensor recalibration and the hassle of resetting alarms or reports. The AssetTag, featuring a QR code, facilitates quick and easy swapping of AiroSensors. When an AiroSensor needs recalibration, simply swap the AiroSensor without any data loss. All pre-set alarms and configurations automatically apply to the new AiroSensor, ensuring a gap-free monitoring experience. With AssetTagging, data loggers can be switched anytime, maintaining continuous and reliable surveillance.
Credits - Five credits are included to register your devices, optionally receive Voice Alerts and purchase calibration certificates if needed.