ContactUsLinkComponent
StarterKit −200 to +200 °C, with AccessPoint, AssetTag and Five Credits
StarterKit −200 to +200 °C, with AccessPoint, AssetTag and Five Credits
# de catalogue 77391-878
Fournisseur:  SENSEANYWHERE BV
New Product
STARTER KIT AIROSENSOR X ER -43 915MHZ
StarterKit −200 to +200 °C, with AccessPoint, AssetTag and Five Credits
# de catalogue 77391-878
Fournisseur:  SENSEANYWHERE BV
New Product
Spécifications
  • Description:
    StarterKit −200...+200 °C, with AccessPoint, AssetTag and 5 credits
Spécifications
A propos de cet article
Explore the SenseAnywhere StarterKit −200 to +200 °C for unparalleled temperature surveillance in extreme environments. Integrating the AiroSensor 20-20-43 and a PT100 Probe, it offers accurate readings for cryogenic applications. Simplified calibration, robust Indoor AccessPoint, and innovative AssetTagging ensure continuous, secure data transmission and seamless sensor management in demanding conditions.
  • Accurate temperature monitoring with both the logger and the PT100
  • 24/7 data for a full audit and always secure access to your data via the Cloud
  • Immediate notification by email, SMS or voice when limits are exceeded
Introducing the SenseAnywhere StarterKit −200 to +200 °C, your ultimate solution for precision temperature surveillance in the most challenging environments. Equipped with the innovative Digital SenseAnywhere Bus (SAB) interface, this AiroSensor seamlessly integrates with our advanced PT100 Probe. The Probe, utilizes smart class A wire-wound ceramic to deliver exceptionally accurate readings over an extensive range from −200 to +200 ˚C. This makes it an ideal choice for monitoring cryogenic applications.
Designed specifically for ultra-low temperature environments, our PT100 Probe ensures accuracy and reliability in conditions as low as –200 °C. The key to its precision lies in the electronics within the connector, which interprets the measured values. This design means that only the PT100 Probe requires calibration, simplifying the process while maintaining the highest measurement standards. Whether you're dealing with cryogenics or other demanding applications, the SenseAnywhere StarterKit −200 to +200 °C is your go-to choice for dependable, precise temperature surveillance.
Indoor AccessPoint - The SenseAnywhere Indoor AccessPoint is the cornerstone of reliable and secure data transmission. It connects our Sensors to the SenseAnywhere Cloud Service, ensuring robust, two-way, fully encrypted communication. Sensors automatically detect the AccessPoint, seamlessly transmitting data to our central database. With advanced protocols for re-transmission and duplicate packet filtering, we guarantee that no event is ever missed. Designed specifically for indoor environments, this AccessPoint is an integral part of our robust surveillance solution.
AssetTagging - AssetTagging by SenseAnywhere revolutionizes sensor calibration and monitoring continuity. This system allows for uninterrupted data logging with interchangeable loggers, eliminating gaps during sensor recalibration and the hassle of resetting alarms or reports. The AssetTag, featuring a QR code, facilitates quick and easy swapping of AiroSensors. When an AiroSensor needs recalibration, simply swap the AiroSensor without any data loss. All pre-set alarms and configurations automatically apply to the new AiroSensor, ensuring a gap-free monitoring experience. With AssetTagging, data loggers can be switched anytime, maintaining continuous and reliable surveillance.
Credits - five credits are included to register your devices, optionally receive Voice Alerts and purchase calibration certificates if needed.
Ordering information: Explore the full range of SenseAnywhere products and services by visiting the Documentation tab, selecting 'Additional Product Details,' and opening the 'Which SenseAnywhere Data Logger Do I Need' document.
Delivery: Begin precise surveillance of your vital assets across extreme temperatures ranging from −200 to +200 °C with our comprehensive StarterKit. This kit includes an AiroSensor 20-20-43, a highly accurate PT100 Probe, an AccessPoint for reliable data transmission, an AssetTag for seamless sensor management, and 5 credits for immediate use.
Customers Who Bought This Also Bought