Economy Constant Temperature Water Bath, 200-230 V, Yamato
# de catalogue 76443-164
Fournisseur: Yamato Scientific
Irregular Voltage
Spécifications
- Description:Digital water bath with Pt100Ω sensor, BM-312A
- Height:240 mm
- Capacity:~5 L
- Electrical:200 - 230 V, 5 - 6 A
- Temperature range:+10 ~ 90 °C
- Weight:~4.5 kg
Spécifications
A propos de cet article
The compact design and simple set-up systems make these water baths easy to use with rotary evaporators.
- 200-230 V model, not for use in USA
- Smaller baths equipped with a drain
- Various sizes available
- Bath protective cover
- Thermal fuse
The larger sizes are equipped with digital setting by UP/DOWN keys.
All models feature thermal fuses for safety.
Caution: 200-230 V model, not for use in USA. Corresponding model with voltage suitable for use in US (76443-162) is also available, please see separate product page.
Customers Who Bought This Also Bought