Specifications
- Description:biOrb humidimist
- Cat. no.:470352-582
Specifications
About this item
Refill your Biorb air mister.
- Specially formulated for Biorb Air and Air 30 models
- Creates a fine mist
- Keeps air clear
- Easy to use
The biOrb humidimist is used specifically with the biOrb AIR 30 and 60 terrariums, ensuring your plants stay healthy and get the humidity they need. Each bottle contains low levels of electrolytes (total dissolved solids) and reduced minerals. Humidimist contains the right level of electrolytes to trigger the sensors inside the misting unit. The ultrasonic mister creates a fine mist that supplies the plants with moisture right down to the roots. Unlike other types of water, humidimist will not produce limescale, clouding, or water spots on the acrylic inside the chamber. Simply pour humidimist into the AIR's misting unit as needed. Includes four, 16.9 fl. oz. bottles. A spray top is also included for manual misting.