ContactUsLinkComponent
Shaken Baby Syndrome Model
Shaken Baby Syndrome Model
# de catalogue 470352-890
Fournisseur:  American 3B Scientific
New Product
Shaken Baby Syndrome Model
# de catalogue 470352-890
Fournisseur:  American 3B Scientific
New Product
Spécifications
  • Description:
    Shaken baby syndrome model
  • Cat. no.:
    470352-890
Spécifications
A propos de cet article
Illustrate the damage caused by SBS.
  • Easy to visualize clear skull
  • Model is 19" long
An infant's vulnerability to shaken baby syndrome (SBS) is powerfully demonstrated with this model. The model's translucent head allows viewers to see how a baby's brain can be harmed. Also emphasizes the importance of supporting an infant's neck. Helps teach prevention of SBS, which can have devastating consequences including permanent brain damage, blindness, and death.
Customers Who Bought This Also Bought