Smokey Sue Model
Fournisseur: American 3B Scientific
New Product
Certificats
A propos de cet article
A classic for teaching the hazards of smoking.
- Two versions available
Smokey sue dramatically demonstrates the quantity of tar collected in the lungs when a single cigarette is smoked. The tar, normally inhaled directly into the lungs, is collected in a transparent tube, and thus shows the quantity of tar which reaches the lungs with each cigarette very clearly. Delivered with stand, 5 collection tubes, and carrying bag. Expand your lesson to include a demonstration stressing the impact of smoking during pregnancy. The smokey sue smokes for two, includes a fetus model showing pollutants that reach the fetus.
Customers Who Bought This Also Bought