Staticide® Dualmat™ ESD Safe Tray Liners, Gray
Fournisseur: ACL Staticide
New Product
Certificats
A propos de cet article
The Dualmat™ 06 Tray liners are durable, static dissipative, and offer reliable static control.
- Two-layer, conductive/dissipative design
- Includes 4 oz. mat cleaner
- Embossed surface
- Made in USA
Mats consist of two layers, with a conductive bottome and dissipative top side. Bottom side resistance measures less than 5×10⁵ ohms. Top side resistance measures 10⁶ to 10⁸ ohms.
Certifications: Meets ANSI/ESDA S20.20.
Customers Who Bought This Also Bought