Staticide® Dualmat™ ESD Mats, Bulk Rolls, Blue
Fournisseur: ACL Staticide
New Product
Certificats
A propos de cet article
The Dualmat™ was developed for companies adhering to electrostatic control and lead-free requirements.
- Top side resistance measures 10⁶ to 10⁸ ohms
- Bottom side resistance measures less than 5×10⁵ ohms
- Embossed surface
- Rolls available in lengths of 40' and widths of 24", 30" and 36"
- Custom cut sizes available
- Mat thickness is 0.15 cm (0.06")
The durable, 2-layer rubber material is inherently dissipative on the top side and conductive on the bottom side. These mats are ideal for electronics manufacturing. Includes 4 oz. mat cleaner.
Certifications: Meets ANSI/ESDA S20.20.
Customers Who Bought This Also Bought