Staticide® Dualmat® ESD Two-Layer Static Dissipative Mats
Supplier: ACL Staticide
New Product
Certificates
About this item
Two-layer static dissipative mat in solid black.
- Top side resistance measures 10⁶ to 10⁸ ohms
- Bottom side resistance measures less than 5×10⁵ ohms
Certifications: Meets ANSI/ESD S20.20.
Customers Who Bought This Also Bought