Eisco Plastic Stirring Rods, Pack of 12
Catalog # 470344-172
Supplier: Wards
Specifications
- Description:Stirring rods
- Cat. no.:470344-172
Specifications
About this item
Stirring rods with rounded ends, made of chemically-resistant polypropylene plastic.
- Pack of 12 multipurpose stirring rods with rounded ends
- Made of high quality, heavy-duty polypropylene
- Autoclavable with very good resistant to chemicals
- Each rod measures 12" in length, and is 0.25" in Ø
Customers Who Bought This Also Bought