Microscope Cover Glasses, Rectangular, No. 1
Fournisseur: FOUR E’S INDUSTRIAL GROUP INC.
New Product
Certificats
A propos de cet article
Four E's premium cover glasses with available selection between super white glass or borosilicate glass 3.3
- Precision-cut for uniform sized pieces
- Ultra-clear glass for optimal visibility
- Premium quality material for durability
- Autoclavable
No. 1 pertains to 0.13 to 0.16 mm thickness. The super white glass offering allows for superior light transmittance for enhanced visibility.
Customers Who Bought This Also Bought