Silane-Coated Adhesive Microscope Slides, Color Frosted
Fournisseur: FOUR E’S INDUSTRIAL GROUP INC.
New Product
Certificats
A propos de cet article
Four E's premium adhesive glass slides with silane coating for superior adhesion.
- Pre-cleaned, ready to use
- Color frosted on one end, one side
- Grounded edges, 45° corners
- Autoclavable
- Colors available: White, green, blue and yellow
The silane coating offers enhanced adhesion performance, specifically designed for histological and plastic sections. It proves effective for routine H&E stains, IHC, ISH, and frozen sections. Compatibility extends to inkjet and thermal transfer printers, as well as permanent markers for seamless marking.
Customers Who Bought This Also Bought