Color Frosted Microscope Slides
Fournisseur: FOUR E’S INDUSTRIAL GROUP INC.
New Product
Certificats
A propos de cet article
Four E's color frosted microscope slides made from premium super white glass for ultra clarity with higher light transmittance.
- Pre-cleaned, ready to use
- Color frosted on one end, one side
- Grounded edges, 45° corners
- Autoclavable
- Colors available: White, green, blue and yellow
The colorfast opaque coating is resistant to commonly used laboratory solvents. The chromatic integrity stays intact even through routine H&E staining.
Customers Who Bought This Also Bought