Plain Microscope Slides
Fournisseur: FOUR E’S INDUSTRIAL GROUP INC.
New Product
Certificats
A propos de cet article
Four E's microscope slides made from high quality soda glass or super white glass.
- Pre-cleaned, ready to use
- Grounded edges, 45° corners
- Autoclavable
Suitable for traditional identification labeling, these plain slides are manufactured using an advanced grinding process to ensure uniform pieces and consistency in flatness and smoothness. The polished edges and corners greatly reduce the risk of scratches and infection during use and handling of the slides.
Customers Who Bought This Also Bought