Nichipet Premium LT Single Channel Pipettes
Fournisseur: Nichiryo America
Certificats
A propos de cet article
The Nichipet Premium LT pipette provides innovation in comfort while providing durability and precise results.
- Mltigates tension to thumb
- Single hand pipetting can be done smoothly to improve operation efficiency
- New design for reducing stroke loading and mitigating fatigue
- Uses an enlarged counter display which is better for visibility and volume setting
- Seal ring for the Premium is tested dispensing/aspirating 600000 times (2 to 1000 μl)
The Nichipet premium LT provides a hyperblowout system improving dispensing efficiency especially with low volume pipetting. It is easily calibrated in lab, and is fully autoclaveable.
Customers Who Bought This Also Bought