ContactUsLinkComponent
Oakton® Flexible Electrode Holder
Oakton® Flexible Electrode Holder
# de catalogue 76221-268
Fournisseur:  Argos Technologies
Flexible Benchtop Electrode Holder
Oakton® Flexible Electrode Holder
# de catalogue 76221-268
Fournisseur:  Argos Technologies
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Flexible benchtop electrode holder, Ø 15.2 cm (6") base
  • Cat. no.:
    76221-268
  • For Use With:
    Up to four standard size electrodes (Ø 14 mm max.) and one ATC probe
Spécifications
A propos de cet article
Keeps electrodes in position.
  • Weighted base pivots 360° for added convenience
Holds up to four standard size electrodes (14 mm dia. max.) and one ATC probe. Weighted base lets you pivot holder 360° for added convenience. Flexible arm lets you move electrode side to side or up and down while keeping electrode at a constant vertical angle.
Customers Who Bought This Also Bought