Safety Pipette Filler, Bulb Type, Bal Supply
Supplier: Tip Industries
Certificates
About this item
This bulb-type safety pipette filler is designed for efficient one-hand operation.
- Made from natural rubber
- Includes 3 glass valves
For use with serological pipettes of varying sizes.
Customers Who Bought This Also Bought