LTS® Filtered Pipette Tips, Rainin® LTS® Compatible
Fournisseur: Azer Scientific Inc.
Certificats
A propos de cet article
Our sterile, 20, 200 and 1000 µl LTS® pippete filter tips are manufactured from super clear high quality polypropylene. The surfaces of the tips are produced through a special process. This process makes the tip inner surface become super hydrophobic, thus significantly reduces sample loss and provides substantially higher reproducibility when working with critical media.
- LTS® compatible
- Low retention for optimal precision and minimal sample loss
- Barrier filter to prevent aerosol contamination
- Certified DNase-/RNase-/pyrogen-/endotoxin-free
Customers Who Bought This Also Bought