Magnaprobe with Magnetic Globe
# de catalogue 470352-176
Fournisseur: United Scientific Supplies
New Product
Spécifications
- Description:Magnaprobe with magnetic globe
- Cat. no.:470352-176
Spécifications
A propos de cet article
Visually demonstrate the magnetic field near the Earth.
- Smooth action magnaprobe
- Detailed globe with countries
- Includes activity guide
The earth acts like it has a huge bar magnet located at its center. This set contains a globe that is 110 mm wide with a magnet contained along its axis. The magnaprobe contains a small magnetthat is allowed to rotate on two axes. This set illustrates the path of the magnetic field along the surface of a globe. The closer to the poles, the steeper the angle of the magnetic field. This is useful in demonstrating the large magnetic field that exists on the earth. The activity guide contains information about the Earth’s magnetic field, Van Allen belts, and the northern lights.
Customers Who Bought This Also Bought