J.T.Baker® BAKERBOND® Q2100 New Generation UHPLC and HPLC Columns
Fournisseur: HICHROM LIMITED
New Product
Certificats
A propos de cet article
BAKERBOND® Q2100 LC phases have been specially designed to solve the challenges of most routine samples. They can also be a good starting point for new method development projects. With options of four widely used chemistries and three particle sizes BAKERBOND® Q2100 can be a useful addition to your analytical toolkit.
- High efficiency UHPLC/HPLC columns
- Based on high purity spherical silica
- C18, C8, phenyl-hexyl and silica phases
- Available with 1.8, 3, 5 µm particle sizes
Customers Who Bought This Also Bought