VWR® Guard Cartridge Holder for J.T.Baker® BAKERBOND® Q2100 UHPLC and HPLC Columns
Fournisseur: HICHROM LIMITED
New Product
Certificats
VWR® Guard Cartridge Holder for J.T.Baker® BAKERBOND® Q2100 UHPLC and HPLC Columns
Fournisseur: HICHROM LIMITED
New Product
A propos de cet article
Customers Who Bought This Also Bought
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
$63.72 to $462.44
SureSTART™ Screw Caps, 9 mm, Level 3 High Performance Applications
Fournisseur: Thermo Scientific
$79.21 to $82.14
WHEATON® ABC Screw Caps, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$101.25 to $1,012.48
WHEATON® MicroLiter Sample Vial Component Kits, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$175.49
VWR® 11 mm Crimp Top Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
$291.56