Miracle® Blue Nitrile Powder-Free Examination Gloves
Fournisseur: High-Tech Conversions
New Product
Certificats
A propos de cet article
Miracle® Blue Nitrile Powder-Free Exam Gloves are 100% rubber latex-free, eliminating the potential risk of rubber latex sensitivities or allergies.
- Excellent 'skin-like' fit and feel
- Ambidestrous
- Single-use only
The Miracle® glove was evaluated by the independent consultants engaged by the Dental Advisor (TDA), and after thousands of uses in their daily practices, the Miracle® glove received a 90% clinical rating. For additional information, see the Miracle® Glove TDA review.
Packaging: Gloves are packaged 200 pieces per box (180 pieces per box for size XL).
Spécifications
- Catalog No:
- 77008-020
- 77007-954
- 77008-058
- 77007-908
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Material:
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile