Bronze Latex Powder-Free Examination Gloves
Fournisseur: High-Tech Conversions
New Product
Certificats
A propos de cet article
Bronze Latex Powder-Free Exam Gloves are a quality choice at an economical and affordable cost.
- Single-use only
- Easy donning and doffing
Though these gloves are made in lighter weight and thinner gauge, they are strong enough for normal everyday examination use in dental, medical, and laboratory applications.
Packaging: Gloves are packaged 100 pieces per box (90 pieces per box for size XL).
Spécifications
- Catalog No:
- 77008-044
- 77007-956
- 77007-870
- 77008-004
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex
- Latex
- Latex
- Latex
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Material:
- Latex
- Latex
- Latex
- Latex
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile