A propos de cet article
VPF Translucent Vinyl Powder-Free Exam Gloves provide a latex-free, powder-free, and economical solution to hand protection.
- Formulated from a special synthetic component and soft PVC material
- Gloves are softer and fit snugly and comfortably to hands
Gloves are 100% rubber latex-free, eliminating the risk of latex protein related allergies.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 77008-064
- 77007-900
- 77007-894
- 77008-008
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Material:
- Vinyl
- Vinyl
- Vinyl
- Vinyl
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile