A propos de cet article
VITEX™ Creamy Beige Synthetic Powder-Free Exam Gloves are made of a special PVC blend. With an exceptional snug fit and some stretchability, they are less baggy than regular vinyl gloves and provide better tactile-sensitivity.
This glove features a creamy beige color that makes them look like latex gloves. They are 100% rubber latex-free, eliminating the risk of latex protein related allergies.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 77008-016
- 77007-916
- 77007-988
- 77008-000
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Material:
- Vinyl
- Vinyl
- Vinyl
- Vinyl
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile