SOCOREX® LAB 45™ Reagent Bottles, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Selection of glass reagent bottles, each supplied with one screw cap.
- Clear bottles are made from borosilicate glass
- Amber bottles are square, round or with jug handle
Perfectly adapted with Calibrex™ bottle top dispensers. Corresponding neck adapters supplied with the dispensers.
Customers Who Bought This Also Bought