Inkjet Label and Overlaminate Kits
Fournisseur: Brady Worldwide
New Product
Certificats
A propos de cet article
Take inkjet printing further and label faster with these perfectly sized label and overlaminate kits.
- Permanent acrylic adhesive
- Matte finish
- Inkjet print-receptive material
- Temperature-resistant, UV-resistant, and water-resistant
- Service temperature: −40 to 140 °F
Just peel, adhere, and apply with these pre-sized labels and overlaminate. Inkjet-printable label is outfitted with a specially formulated topcoat that invigorates colors and label appearance. Label topcoat also protects against smudges, smears, and splotches. Acrylic overlaminate provides up to 10 years of outdoor durability through improved UV and abrasion resistance. Overlaminate is oversized by a quarter inch for easier application and complete coverage.
An ink cartridge is required to print label material; use part number J20-CMY for J2000 printer, J40-CMY for the J4000 printer, or J50-CMYK for the J5000 printer.
Customers Who Bought This Also Bought