Self-laminating Vinyl Wrap-Around Labels
Fournisseur: Brady Worldwide
New Product
Certificats
A propos de cet article
Use high-performance, self-laminating labels to identify wires and cables. Bulk label rolls are ideal for continuous printing, reducing the frequency of label changeouts. Wraptor™ A6200 Bulk label roll adapter required to utilize bulk label rolls.
- Permanent acrylic adhesive
- Matte finish
- Flame-retardant, self-laminating material
- Chemical-resistant, humidity-resistant, oil-resistant, water-resistant, and weather-resistant
- Service temperature: −94 to 158 °F
- UL approved
Protect printed text using a self-laminating format. Labels feature a white printable zone with translucent overlaminating area. Labels are self-extinguishing and resist chemicals, humidity, oil, and water. Highly conformable with a durable, permanent acrylic adhesive for indoor/outdoor use.
Use with Wraptor™ A6200 Wrap printer applicator and Wraptor™ A6200 bulk label roll adapter (required). A ribbon is required to print this material; use part number R4300.
Customers Who Bought This Also Bought