Inkjet Vinyl Circular Labels
Fournisseur: Brady Worldwide
New Product
Certificats
A propos de cet article
Make an impact with this inkjet-printable vinyl label outfitted with a specially formulated topcoat that invigorates colors and label appearance.
- Permanent acrylic adhesive
- Matte finish
- Inkjet print-receptive material
- Chemical-resistant, temperature-resistant, UV-resistant, and water-resistant
- Service temperature: −40 to 140 °F
Ideal for circular machine gauge labels common to Lean/5S applications. Flexible material withstands harsh elements including liquids, alcohol, chemicals, and indoor fluorescent and UV lighting. Topcoat also protects against smudges, smears, and splotches.
An ink cartridge is required to print this material; use part number J20-CMY.
Customers Who Bought This Also Bought