Clear Acrylic Overlaminate for BradyJet™ Labels
Fournisseur: Brady Worldwide
New Product
Certificats
A propos de cet article
Achieve up to 10 years of outdoor durability through improved UV and abrasion resistance with acrylic overlaminate for BradyJet™ labels.
- Permanent acrylic adhesive
- Gloss finish
- Optically clear material
- Service temperature: −20 to 180 °F
No need to cut to size, simply peel and apply with these pre-sized rolls that are perfectly sized to their corresponding BradyJet™ labels. Overlaminate is a quarter inch wider than its corresponding BradyJet™ label to ensure complete coverage.
Customers Who Bought This Also Bought