ContactUsLinkComponent
Grip Strength Tests
Grip Strength Tests
# de catalogue 76670-290
Fournisseur:  MAZE ENGINEERS
Grip Strength Tests
# de catalogue 76670-290
Fournisseur:  MAZE ENGINEERS
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Grip strength test, mouse/rat
Spécifications
A propos de cet article
The Grip Strength Test was developed in the 1970s (Meyer et al., 1979; Cabe et al., 1978) and is a widely used apparatus to evaluate motor function in rodents such as rats or mice.
  • Gripping plate area; mouse: 20×20 cm, rat: 25×25 cm
  • Stainless steel, detachable
The Grip Strength Test apparatus uses both small and large grip plates to match specific rodent models. The apparatus measures tensile force and easily outputs mean values. The instrument has an error value elimination function, which can eliminate operational errors in time, eliminate false values, ​​and improve the authenticity of data. The instrument can read instrument data online with a PC. This apparatus can be used as either a forelimb grip strength test or a hindlimb grip strength test.
Customers Who Bought This Also Bought